Description
Available in a variety of colors, vented or unvented
- Cap style hard hats constructed of HDPE
- Four-point ratchet suspension
- Uvicator lets wearer known when to replace hard hat due to UV exposure
- Suspension sits lower on head to reduce pressure and increase security
- Smooth, easy turning ratchet suspension
- Dielectric: Class C, G, and E
- 3M Catalog# H-701R-UV, H-701V-UV, H-702R-UV, H-702V-UV, H-703R-UV, H-703V-UV, H-704R-UV, H-704V-UV, H-709R-UV, H-711R-UV, H-711V-UV, H-712V-UV, H-713R-UV, H-713V-UV, H-712R-UV
Compliance
Meets requirements of ANSI/ISEA Z89.1-2009 Type 1, Class C.
Specifications
Specifications
| Color | White |
| Includes | Uvicator |
| Model | H-701R-UV |
| Product Type | Hard Hat with Uvicator |
| Suspension Type | 4-Point Ratchet |
| Ventilated | Not Ventilated |