Learn More
3M™ Hard Hat with Uvicator™ Sensor
Catalog No. 17623788 Shop All 3M Products
Change view
Click to view available options
Color:
Black
Blue
Bright Yellow
Green
Pink
Tan
White
Yellow
Ventilated:
Not Ventilated
Ventilated
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Ventilated
17-623-788 Tan Ventilated
17-623-634 White Not Ventilated
17-623-636 Yellow Not Ventilated
17-623-635 White Ventilated
17-623-638 Blue Not Ventilated
17-623-637 Yellow Ventilated
17-623-639 Blue Ventilated
17-623-640 Green Not Ventilated
17-623-641 Green Ventilated
17-623-642 Bright Yellow Not Ventilated
17-623-786 Tan Not Ventilated
17-623-793 Black Ventilated
17-623-796 Pink Not Ventilated
17-623-798 Pink Ventilated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Catalog No. 17-623-788 Supplier 3M™ Supplier No. 7100075595
Protective hard hat with vents, comfortable 4-point ratchet suspension and standard brow pad
Available in a variety of colors, vented or unvented
  • Cap style hard hats constructed of HDPE
  • Four-point ratchet suspension
  • Uvicator lets wearer known when to replace hard hat due to UV exposure
  • Suspension sits lower on head to reduce pressure and increase security
  • Smooth, easy turning ratchet suspension
  • Dielectric: Class C, G, and E
  • 3M Catalog# H-701R-UV, H-701V-UV, H-702R-UV, H-702V-UV, H-703R-UV, H-703V-UV, H-704R-UV, H-704V-UV, H-709R-UV, H-711R-UV, H-711V-UV, H-712V-UV, H-713R-UV, H-713V-UV, H-712R-UV
Compliance
Meets requirements of ANSI/ISEA Z89.1-2009 Type 1, Class C.
TRUSTED_SUSTAINABILITY
Specifications
Color Tan
Model H-711V-UV
Product Type Hard Hat with Uvicator
Ventilated Ventilated