Learn More
3M™ Lexa™ Safety Eyewear
Catalog No. 19033439S Shop All 3M Products
Change view
Click to view available options
Lens Tint:
Clear
Gray
Frame Color:
Black
Metallic Slate
Slate
Lens Coating:
Anti-fog
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint Frame Color Lens Coating
19-033-439S Clear Slate -
19-033-440S Gray Slate Anti-fog
19-034-443 Clear Metallic Slate -
19-150-0143 Clear Black -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Catalog No. 19-033-439S Supplier 3M™ Supplier No. 7010045700
This item is not returnable. View return policy
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
No need for browguards or sideshields
Combine extra-wide viewing range and contoured design that follows facial outline; eliminates the need for bulky browguards and distracting sideshields.
  • Lightweight, full-coverage eye protection
  • DX™ anti-fog hard coat protects lens from fog, scratching, and most chemicals
  • Lens are easy to switch — simply snap off the temples and re-attach them to the desired lens
  • Fully customizable with three lens angles and four temple lengths to fit virtually everyone
  • Slate temples
  • 3M Catalog# 15100-00000-20 , 3M Catalog# 15200-00000-20 , 3M Catalog# 15204-00000-20
Compliance
Meets the requirements of ANSI™ Z87.1-2010 and CSA Z94.3-2007.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Meets the requirements of ANSI Z87.1-2010 and CSA Z94.3-2007.
Lens Tint Clear
Frame Color Slate
Model 15200-00000-20
Lens Material Polycarbonate