Description
Combine extra-wide viewing range and contoured design that follows facial outline; eliminates the need for bulky browguards and distracting sideshields.
- Lightweight, full-coverage eye protection
- DX™ anti-fog hard coat protects lens from fog, scratching, and most chemicals
- Lens are easy to switch — simply snap off the temples and re-attach them to the desired lens
- Fully customizable with three lens angles and four temple lengths to fit virtually everyone
- Slate temples
- 3M Catalog# 15100-00000-20 , 3M Catalog# 15200-00000-20 , 3M Catalog# 15204-00000-20
Compliance
Meets the requirements of ANSI™ Z87.1-2010 and CSA Z94.3-2007.
Specifications
Specifications
| Lens Coating | Anti-fog |
| Certifications/Compliance | Meets the requirements of ANSI Z87.1-2010 and CSA Z94.3-2007. |
| Lens Tint | Gray |
| Frame Color | Slate |
| Model | 15204-00000-20 |
| Lens Material | Polycarbonate |