Description
Includes
Light Vision and Light Vision 2 glasses supplied with lanyard and storage bag. Light Vision OTG supplied with storage bag.
- Dual hands-free temple LED lights swivel and adjust for bright directional beams
- Wraparound design provides wide coverage to protect against particulates
- Impact-resistant, clear polycarbonate lens with antifog coating absorbs 99.9% UV
- Soft temples and nose pads for extended wear comfort
- Light Vision 2 glasses feature adjustable temple lengths
- Over-the-glass style fits over prescription glasses; features adjustable length temples and Secure-Grip™ temple tips
- Light Vision 2 available with bifocal readers in three strengths for workers who have difficulty reading small print or are engaged in small detail work
- Batteries provide up to 50 hours of continuous use
- 3M Catalog# 11356-00000-10, 11476-00000-10, 11477-00000-10, 11478-00000-10, 11479-00000-10
Compliance
Meets ANSI™ Z87.1-2010 standards. LightVision 2 also meets CSA Z94.3-2007 standards.
Specifications
Specifications
| Certifications/Compliance | Meets ANSI Z87.1-2010 standards. LightVision 2 also meets CSA Z94.3-2007 standards. |
| Lens Tint | Clear |
| Frame Color | Gray |
| Includes | Light Vision |
| Magnification Power | 0 |
| Temple Color | Black |
| Lens Material | Polycarbonate |