Learn More
3M™ Metaliks™ Eyewear
Catalog No. 19051436 Shop All 3M Products
Change view
Click to view available options
Frame Color:
Brushed Nickel
Polished Metal
Silver
Lens Tint:
Clear
Gray
Indoor/Outdoor Mirror
Lens Coating:
Anti-fog
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint Lens Coating
19-051-436 Silver Indoor/Outdoor Mirror -
19-051-434 Polished Metal Clear Anti-fog
19-121-271 Brushed Nickel Clear Anti-fog
19-121-272 Brushed Nickel Gray Anti-fog
19-121-273 Brushed Nickel Indoor/Outdoor Mirror -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Catalog No. 19-051-436 Supplier 3M™ Supplier No. 7000127571
This item is not returnable. View return policy
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Offers distinctive style, maximum coverage, and adjustable comfort
  • Impact-resistant polycarbonate lenses with antifog hardcoat† to resist fog, scratches, and most chemicals
  • Soft, adjustable nose bridge and temple tips
    • Standard Metaliks feature a 10-base de-centered lens for optimum front and side protection
    • Metaliks Sports feature brushed nickel frame with athletic-shaped lens design
    • Metaliks GT feature a race-inspired design with a bronze frame
    • 3M Catalog# 11343-10000-20, 11344-10000-20, 11345-10000-20, 11554-00000-20, 11555-00000-20, 11556-00000-20, 15170-10000-20, 15172-10000-20
Compliance
Meets the requirements of ANSI™ Z87.1-2010 and CSA Z94.3-2007.
†I/O Mirror lens have hardcoat only, not antifog hardcoat.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Meets the requirements of ANSI Z87.1-2010 and CSA Z94.3-2007.
Lens Tint Indoor/Outdoor Mirror
Frame Color Silver
Model 15172-10000-20