Learn More
Boston Bioproducts Inc Acetate Buffer (3 M, pH 5.2) - 250ml
Supplier:  Boston Bioproducts Inc BM225250ML
Encompass
The Acetate Buffer is commonly used to maintain pH within 3.6 and 5.6, making it ideal for processes requiring a stable environment at this pH. It is effective in applications including the purification and precipitation of nucleic acids, protein crystallization, and preparing staining solution for gels after protein gel electrophoresis. At a 3M concentration, this buffer has a strong buffering capacity due to an abundance of acetic acid to acetate ions.
Catalog No. NC9823599
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.