Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Nitro™ Ear Plugs

Catalog No. 191300465 Shop All 3M Products
Change view
Click to view available options
Corded:
Uncorded
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Corded
19-130-0465 Uncorded
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-130-0465 Supplier 3M™ Supplier No. 7000127250

NRR: 32 dB

  • Ultra-soft foam formulation for all-day comfort
  • Tapered shape for easy insertion and canal-hugging seal
  • Four vibrant colors in each box: orange, red, pink, and purple
  • 3M Catalog# P1000 , 3M Catalog# P1001

Compliance

3M™ ear plugs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.

TRUSTED_SUSTAINABILITY

Specifications

Color Multi-color
Certifications/Compliance CSA Class AL
Size Regular
For Use With (Application) For easy insertion and canal-hugging seal
Model P1000
Noise Reduction Rating (NRR) 32 dB
Corded Uncorded
Plug Shape Tapered
Product Type Nitro™ Earplug
Material Polyurethane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.