Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Nitro™ Tattoo™ Ear Plugs

Catalog No. 191304170 Shop All 3M Products
Change view
Click to view available options
Uncorded
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-130-4170 Uncorded
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-130-4170 Supplier 3M™ Supplier No. 7000127190
This item is not returnable. View return policy

NRR: 32dB

  • Tapered shape for all-day comfort
  • Feature barbed wire “tattoo” graphic
  • Bright neon green for high visibility
  • 3M Catalog# P1100

Compliance

3M™ ear plugs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.

TRUSTED_SUSTAINABILITY

Specifications

Color Green
Certifications/Compliance CSA Class AL
Size Regular
For Use With (Application) Designed for style-conscious workers
Noise Reduction Rating (NRR) 32 dB
Corded Uncorded
Plug Shape Tapered
Product Type Nitro™ Tattoo™ Earplug
Material Polyurethane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.