Learn More
3M™ OX™ Eyewear
Catalog No. 19153644 Shop All 3M Products
Change view
Click to view available options
Lens Coating:
Anti-fog
Uncoated
Temple Style:
Secure-Tip
Spatula
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Coating Temple Style
19-153-644 Uncoated Spatula
19-121-306 Anti-fog Spatula
19-153-645 Anti-fog Secure-Tip
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Catalog No. 19-153-644 Supplier 3M™ Supplier No. 7000002372
This item is not returnable. View return policy
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Designed to fit over prescription glasses
  • Seamless Duralite polycarbonate lens with built-in sideshields provides an uninterrupted sight range and optically correct peripheral vision
  • Clear lens provides 99.9% UV protection
  • Optional DX™ antifog hardcoat resists fogging, scratching, static and chemicals
  • Choice of standard spatula temples or adjustable Secure-Grip™ temples with a pivoting axis for a secure, custom fit
  • 3M Catalog# 12159-00000-20 , 3M Catalog# 12163-00000-20 , 3M Catalog# 12166-00000-20
Compliance
Meets ANSI Z87.1-2015 a nd CSA Z94.3 standards.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Uncoated
Certifications/Compliance ANSI Z87.1-2015, CSA Z94.3
Lens Tint Clear
Frame Color Black
Model 12159-00000-20
Temple Style Spatula
Lens Material Duralite Polycarbonate