Learn More
3M™ Particulate Filter 2071, P95 Respiratory Protection
Catalog No. 19002714 Shop All 3M Products
Change view
Click to view available options
P95 particulate
Catalog No.
19-002-714 P95 particulate
1 options
This item is not returnable. View return policy
Catalog No. 19-002-714 Supplier 3M™ Supplier No. 7000002058
Add to Cart
Add to Cart
This item is not returnable. View return policy
Requires
Adapter Cat. No. 17-459-4, if used with 5000 Series Respirators or as a prefilter for 6000 Series Cartridges.
For use with 3M Reusable Respirators
Designed for use with 3M™ 5000 Series Respirators or 6000 Series cartridges with 3M Adapter 502, or 6000, 7000 or FF-400 Series facepieces with bayonet filter holders.
P95 Particulate
  • 95% efficient against solid and liquid aerosols, including oils
  • Applications such as grinding, sanding, sawing, sweeping and bagging
  • Can also be used as a prefilter in certain paint spraying and pesticide applications
  • 3M Catalog# 2071
These safety products must be used in accordance with NIOSH approvals and OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.
TRUSTED_SUSTAINABILITY
Specifications
Cartridge Type Filter Cartridge
Certifications/Compliance NIOSH
Product Type P95 Particulate Filter
For Use With (Application) Cleaning, demolition, facility maintenance, grinding, painting, sanding, welding
For Use With (Equipment) 3M™ reusable respirator
Model 2071
Length (English) 4.3 in.
Length (Metric) 10.9 cm
Height (English) 4.3 in.
Height (Metric) 10.9 cm