Description
- Easily snaps into employee's own ear muffs (sold separately) to enable fit test
- For use with 3M™ PELTOR™ X Series ear muffs and the E-A-Rfit™ dual-ear validation system
- Probed cushions can be cleaned with hygiene wipes and reused
- Adapter clips hold microphones next to ear muffs during testing
Compliance
3M™ products must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Specifications
Specifications
| Cushion Color | Black |
| For Use With (Equipment) | X Series X3 Ear Muffs |
| For Use With (Application) | Fit testing with the 3M™ E-A-Rfit™ dual ear validation system |
| Model | 393-3003-2 |
| Product Type | Probed Ear Muff Test Cushions |
| Material | PVC |