Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ PELTOR™ External PTT for WS™ LiteCom Pro III Headset

Catalog No. 19072320 Shop All 3M Products
Change view
Click to view available options
Accessory LiteCom III
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-072-320 Accessory LiteCom III
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-072-320 Supplier 3M™ Supplier No. 7000039666

PELTOR™ External PTT LiteCom Pro III IS Headset includes a Push-to-Talk (PTT) button for convenient positioning and activation.

  • Conveniently position the PTT button for easy activation
  • Certified to be intrinsically safe for use in potentially explosive atmospheres by CSA Group
  • For use with the WS LiteCom Pro III Headsets

Compliance

3M™ ear muffs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.

TRUSTED_SUSTAINABILITY

Specifications

Color Black
For Use With (Equipment) WS™ LiteCom Pro III Headsets
For Use With (Application) Push-to-Talk (PTT) adapter with 4-pin XLR plug
Model FL5602-50
Pairs per Package 1 Ea.
Product Type PELTOR
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.