Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ PELTOR™ Probed Electronic Ear Plugs

Catalog No. 19072248 Shop All 3M Products
Change view
Click to view available options
Size:
Large
Regular
Small
Universal
Plug Color:
Brown
Gray
Tan
Plug Shape:
Cone
Flanged
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Plug Color Plug Shape
19-072-248 Large Brown Flanged
19-072-287 Small Brown Flanged
19-072-265 Regular Tan Flanged
19-072-273 Large Gray Flanged
19-072-308 Universal Gray Cone
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19-072-248 Supplier 3M™ Supplier No. 7100225706
Add to Cart
Add to Cart

PELTOR™ Probed Electronic ear plugs are designed to be used with the 3M™ E-A-Rfit™ Dual ear Validation System and enables you to measure a Personal Attenuation Rating (PAR) for the 3M PELTOR TEP, LEP, and EEP Electronic ear plugs. The Probed Electronic ear plugs are for testing purposes only and are NOT to be used for hearing protection.

  • Test ear pieces are modified with a probe tube for use during fit testing with the 3M™ E-A-Rfit™ Dual-ear Validation System
  • Contains 10 probed ear pieces and 50 pair of ear tips per case
  • 3M PSD products are occupational use only

Compliance

3M™ ear plugs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.

TRUSTED_SUSTAINABILITY

Specifications

Size Large
For Use With (Application) Health and Safety Testing
High Visibility No
Model 7100225706
Packaging Type Poly bag
Pairs per Package 50 Pairs/Cs
Plug Material ABS, elastomeric polymer
Plug Color Brown
Corded Uncorded
Plug Shape Flanged
Disposable Reusable
Metal Detectable No
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.