Learn More
Supply Solutions 3M - Petrifilm Count Plates. Type: Count Plate; Incubation: 24 Hours; For Use With: Coliform.; Type: Count Plate; Incubation: 24 Hours
Supplier: Supply Solutions 3M 6404
            3M Petrifilm E.coli/Coliform Count Plate identifies both E. coli and other coliforms with confirmed results within 24-48 hours. By eliminating the confirmation steps you increase productivity and reduce overall lab costs. For achieving fast, accurate results in 24 or 48 hours, follow these three easy steps: 1) Inoculate - easy to inoculate, lift the top film and add sample 2) Incubate - designed to maximize incubator space 3) Enumerate - confirmed coliforms are red and blue colonies with associated gas bubbles. Confirmed E.coli coliforms are blue colonies with associated gas bubbles. 3M Petrifilm Plates can also be read in 4 seconds using the 3M Petrifilm Plate Reader.; Type: Count Plate; Incubation: 24 hours; Min. Population: -; Shelf Life: 1 year after open; For Use With: Escherichia coli and coliform enumeration; Plates are sample-ready, eliminating the time-consuming, cumbersome step of preparing media/agar dishes; 3M Petrifilm Plates use 75% less energy, use nearly 80% less wat
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.