Learn More
3M™ Polycarbonate Faceshields
Catalog No. 19040054 Shop All 3M Products
Change view
Click to view available options
Lens Tint:
Clear
Gold-plated, Clear, Gold-plated
Gold-plated, Dark Green, Gold-plated
Medium Green
Medium Green, Gold-plated
Shade 3
Shade 5
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
19-040-054 Medium Green, Gold-plated
19-040-047 Shade 5
19-040-051 Clear
19-040-055 Gold-plated, Dark Green, Gold-plated
19-040-053 Gold-plated, Clear, Gold-plated
19-040-044 Shade 3
19-040-052 Medium Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 19-040-054 Supplier 3M™ Supplier No. 7000144939
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes
Faceshield only. (Head gear is sold separately.)
Protects workers from infrared hazard exposure.
  • 3M Total Performance faceshields feature dual-coat technology that provides a durable, abrasion-resistant hard coat on the outside and an anti-fog coating on the inside. Made of durable, splash and impact-resistant polycarbonate.
  • 3M provides reliable head and face protection products that meet regulated performance and safety requirements.
  • 3M Catalog# 82600-00000, 82601-00000, 82602-00000, 82603-00000, 82604-00000, 82705-10000, 82706-10000
  • For use with 3M H-Series Hard Hats/Headgear (H24M/7000002292, H24T/7100113761, H18/7000002291, H18-S/7000052752, H8A/7000002290, H10/7000052751, H8A-2/7000127237).
Compliance
Meets high-impact requirements of ANSI Z87.1-2010
TRUSTED_SUSTAINABILITY
Specifications
Thickness (English) 0.75 in.
Lens Tint Medium Green, Gold-plated
For Use With (Equipment) 3M H-Series Hard Hats/Headgear
Model 82603-00000
Lens Material Polycarbonate