Description
- Lens and temple tip color options
- Soft-tipped temples
- Soft comfort nose bridge
- Durable polycarbonate lenses absorb 99.9% UV
- 3M Catalog# 12261-00000-20, 12263-00000-20, 12264-00000-20, 12265-00000-20, 12266-00000-20, 12268-00000-20
Compliance
Meets ANSI™ Z87.1-2010 and CSA Z94.3 standards.
Specifications
Specifications
| Certifications/Compliance | Meets ANSI Z87.1-2010 and CSA Z94.3 standards. |
| Lens Tint | Indoor/Outdoor Mirror |
| Frame Color | Silver |
| Model | 12268-00000-20 |
| Temple Color | Red |
| Lens Material | Polycarbonate |