Learn More
3M™ H-700 Series Cap Style Hard Hats with 4-Point Pinlock Suspension
Catalog No. 17623311 Shop All 3M Products
Change view
Click to view available options
Color:
Black
Blue
Bright Orange
Bright Yellow
Gray
Green
Navy Blue
Orange
Pink
Red
Tan
White
Yellow
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-623-311 Orange
17-623-310 Red
17-623-309 Yellow
17-623-308 White
17-623-328 Navy Blue
17-623-329 Bright Yellow
17-623-330 Gray
17-623-331 Bright Orange
17-623-332 Green
17-623-333 Blue
17-623-784 Tan
17-623-789 Black
17-623-794 Pink
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 17-623-311 Supplier 3M™ Supplier No. 7000030061
Protects top of the head due to potential hazard of small falling objects or exposed fixed objects
  • Easy to adjust suspension offers comfort
  • Standard brow pad enhances comfort and extends shelf life
  • Meets requirements of ANSI/ISEA Z89.1-2009 Type 1, Class C, G and E
  • Short brim for upward visibility
  • Slots allow for attachment of safety accessories
  • Available in various colors
  • 3M Catalog# H-701P, H-702P, H-703P, H-704P, H-705P, H-706P, H-707P, H-708P, H-709P, H-710P, H-711P, H-712P, H-713P
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Meets ANSI/ISEA Z89.1-2009 Type 1, Class C, G and E
Color Orange
Includes Standard brow pad
Model H-706P
Product Type Cap-style Hard Hat
Quantity 20/Cs.
Suspension Type 4-Point Pin-Lock