Learn More
3M™ H-700 Series Cap Style Hard Hats with 4-Point Pinlock Suspension
Protects top of the head due to potential hazard of small falling objects or exposed fixed objects
Supplier:  3M™ 7000127789
 View more versions of this product
Catalog No. 17-623-329
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
  • Easy to adjust suspension offers comfort
  • Standard brow pad enhances comfort and extends shelf life
  • Meets requirements of ANSI/ISEA Z89.1-2009 Type 1, Class C, G and E
  • Short brim for upward visibility
  • Slots allow for attachment of safety accessories
  • Available in various colors
  • 3M Catalog# H-701P, H-702P, H-703P, H-704P, H-705P, H-706P, H-707P, H-708P, H-709P, H-710P, H-711P, H-712P, H-713P
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Meets ANSI/ISEA Z89.1-2009 Type 1, Class C, G and E
Standard brow pad
Cap-style Hard Hat
4-Point Pin-Lock
Bright Yellow
H-709P
20/Cs.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.