Learn More
3M™ QX™ Safety Glasses
Catalog No. 19153630 Shop All 3M Products
Change view
Click to view available options
Lens Tint:
Clear
Gray
Product Type:
QX 1000
QX 2000
QX 3000
Temple Style:
Adjustable
Sport Grip
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint Product Type Temple Style
19-153-630 Clear QX 1000 Adjustable
19-153-636 Clear QX 3000 Sport Grip
19-153-631 Gray QX 2000 Adjustable
19-153-632 Clear QX 2000 Adjustable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Catalog No. 19-153-630 Supplier 3M™ Supplier No. 7000052813
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Seamless lens for distortion-free vision
  • Sleek wraparound lens provides a wide field of vision and excellent side protection
  • Coated polycarbonate lens resists fogging, scratching, static and chemicals; provides 99.9% UV protection
  • Adjustable temples with changeable lengths
  • Lens is easily replaceable using the Easy Change™ system
  • Black frames
  • 3M Catalog# 12101-10000-20, 12115-10000-20, 12109-10000-20, 12100-10000-20
Sport Grip™ temples fit under headgear and ear muffs
Compliance
Meet ANSI™ Z87.1-2010 and CSA Z94.3 standards.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Meet ANSI Z87.1-2010 and CSA Z94.3-2007 standards.
Lens Tint Clear
Product Type QX 1000
Frame Color Black
Temple Style Adjustable
Lens Material Polycarbonate