Learn More
3M™ QX™ Safety Glasses
Seamless lens for distortion-free vision
Supplier:  3M™ 7000127528
 View more versions of this product
Catalog No. 19-153-631
Alert:
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
This item is not returnable. View return policy
Description
Description
  • Sleek wraparound lens provides a wide field of vision and excellent side protection
  • Coated polycarbonate lens resists fogging, scratching, static and chemicals; provides 99.9% UV protection
  • Adjustable temples with changeable lengths
  • Lens is easily replaceable using the Easy Change™ system
  • Black frames
  • 3M Catalog# 12101-10000-20, 12115-10000-20, 12109-10000-20, 12100-10000-20
Sport Grip™ temples fit under headgear and ear muffs
Compliance
Meet ANSI™ Z87.1-2010 and CSA Z94.3 standards.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Meet ANSI Z87.1-2010 and CSA Z94.3-2007 standards.
QX 2000
Adjustable
Gray
Black
Polycarbonate
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.