Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Replacement Explosive Sensor

Catalog No. 19805694 Shop All 3M/Aerion Products
Change view
Click to view available options
Components Measured:
LEL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured
19-805-694 LEL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-805-694 Supplier 3M™ Supplier No. 931000000/DEL

missing translation for 'validMainframeMsg'

For 250/350 series detector.

  • For monitoring %LEL
  • Quantity: 1/Cs.

Specifications

Components Measured LEL
For Use With (Equipment) 250/350 Series Detector
Type Replacement Explosive Sensor
Sensor Types Explosive Sensor
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.