Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ H-Series Sealed Seam Respirator Hood

Catalog No. 19121804 Shop All 3M Products
Change view
Click to view available options
Sealed-seam respirator hood
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-121-804 Sealed-seam respirator hood
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-121-804 Supplier 3M™ Supplier No. 7000126317

Replacement hood for use with H-Series respirators.

  • These safety products must be used in accordance with the user instructions, warnings and limitations accompanying each product.
  • 3M Catalog# H-610-5

TRUSTED_SUSTAINABILITY

Specifications

Assigned Protection Factor (APF) 1000
Certifications/Compliance OSHA
Color White
Material Fabric
Product Type Respirator Hood
Size Standard
For Use With (Application) Assembly, machine operation
For Use With (Equipment) Belt mounted powered air purifying respirator or supplied air component
Includes Collar
Model H-610-5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.