Learn More
3M™ Responder Filter Media Canister
NIOSH-approved for respiratory protection against CN, CS, and as a P100 particulate filter. May be used for exposure to tear gases and pepper spray during riot and crowd control operations.
Supplier: 3M™ 7000131702
Description
- NIOSH-approved with 3M™ Full Facepiece 6700DIN, 6800DIN, 6900DIN, 3M™ Full Facepiece 7800S. and 3M™ Full Facepiece FR-M40 and FR-7800B.
- 3M Catalog# 450-02-11R06
Specifications
Canister Cartridge | |
Black/Magenta | |
Canister Respirator | |
3M™ full facepiece 6700din, 6800din, 6900din, 7800s, fr-m40, fr-7800b | |
6/Pk. |
NIOSH | |
P100 | |
Respiratory protection against CN, CS, tear gas and pepper spray | |
450-02-11R06 |