Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Responder Filter Media Cartridge

Catalog No. 19813754 Shop All 3M Products
Change view
Click to view available options
Quantity:
4/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-813-754 4/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-813-754 Supplier 3M™ Supplier No. 7000126509
Add to Cart
Add to Cart

Filters tear gases and pepper spray, organic vapor, H2S, formaldehyde and acid gas

FR-64 Cartridge
NIOSH-approved for protection from organic vapors, acid gases, ammonia/methylamine, formaldehyde, hydrogen sulfide, phosphine, o-chlorobenzylidene malononitrile (CS) and α-chloroacetophenone (CN), and as a P100 filter

Manufacturer advises that spent cartridges and canisters be disposed of in accordance with applicable federal, state, and local laws and regulations. These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.

  • 3M Catalog# FR-64

Order Info

Technical Data Bulletins are available describing the performance of these filter media against the listed challenge agents. For more information, contact your Fisher Safety Customer Service Representative at 1-800-772-6733.

TRUSTED_SUSTAINABILITY

Specifications

Cartridge Type Respirator Cartridge
Certifications/Compliance MIL-PRF-51560A, EA-C-1704, NIOSH
Connection Type Single DIN threaded
Color Black
Product Type Fr-64 Canister Respirator
For Use With (Application) To filter chemical warfare agents, many industrial chemicals and particulates
For Use With (Equipment) 3M™ full facepiece fr-m40, 6000, 7800s
Model FR-64
Quantity 4/Cs.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.