Learn More
3M™ S-Series Premium Suspension Hoods and Replacements
Lightweight, loose-fitting hoods and head Covers offer both comfort and protection
Manufacturer:  3M™ 7000127428
 View more versions of this product
Catalog No. 19-805-610
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Integrated Suspension Hoods and Head Covers
  • Head covers provide head and face coverage; hoods cover head, face, and neck
  • Fully assembled and ready to use
  • Suitable for applications that require frequent replacement of the entire headgear assembly, such as pharmaceutical manufacturing, or situtations where there is less soiling of the outer fabric
  • Integrated models feature wide faceseal with soft elastic for improved comfort and reduced noise
  • Premium Reusable Suspensions with Hoods
    • Ideal for high disposal rate environments
    • Soiled hood is replaced; suspension and air ducting are reused
    • Rigid ducting for consistent air path
    • Externally adjustable airflow control allows wearer to modify airflow distribution
    • Suspension features independent adjustments for the crown and head circumference; allows custom fit for wide range of head sizes
    • 3M Catalog# S-133L-5, S-133S-5, S-433S-5, S-533L, S-533S, S-707-10, S-757, S-805-5, S-807-5, S-857
Specifications
Specifications
Polypropylene
White
Headcover
Assembly, facility maintenance, machine operation, painting, sanding
9.06 in.
Head and face coverage. Integrated Comfort suspension. General purpose fabric.
50.49 cm
5/Cs.
27 cm
NIOSH
19.88 x 10.63 x 9.06 in. (50.49 x 27 x 23.01 cm)
Medium/Large
3M™ powered air purifying respirator and supplied air respirator valves
23.01 cm
19.88 in.
S-133L-5
10.63 in.