Description
- Strong polypropylene tape film backing with rubber resin adhesive
- Heavy-duty, conformable backing with higher edge tear and split resistance.
- Performs well on single- and double-wall corrugated cartons with higher value contents and medium-weight regular slotted containers
- Best when applied at room temperature
- 3 in. core
- 3M Catalog# , 373, 373, 373, 373, 373, 373, 373
Specifications
Specifications
| Model | 373 |