Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ SecureFit™ 400 Series Readers Protective Eyewear

Catalog No. 17623770 Shop All 3M Products
Change view
Click to view available options
Magnification Power:
+1.50
+2.00
+2.50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Magnification Power
17-623-770 +2.00
17-623-769 +1.50
17-623-771 +2.50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 17-623-770 Supplier 3M™ Supplier No. 7100115325
Add to Cart
Add to Cart
This item is not returnable. View return policy

Features reading diopter to help workers who have difficulty reading small print or who are engaged in small detail work

Provides comfortable, durable, lightweight eye protection.

  • Clear anti-fog coated polycarbonate lens absorb 99.9% UV
  • Available with +1.5, +2.0, or +2.5 reading diopters
  • Pressure Diffusion Technology allows temples to naturally adjust to individual head sizes eliminating need to stock multiple frame sizes at worksites
  • Soft rubber nose bridge
  • Wide viewing area
  • Rubber temple tips
  • Color-accented temples
  • Frameless
  • Z87+ lens marking
  • One size fits all
  • 3M Catalog# SF415AF , 3M Catalog# SF420AF , 3M Catalog# SF425AF

Applications:

Facility Maintenance, Sawing, Masonary, Machining, Electrical, Machine Operations, Drilling, Assembly, Painting, Cleaning, Grinding, Pouring/Casting, Remediation, Sanding, Welding, Demolition, Riveting, Chipping, UV Protection, Power Fastening

Compliance

Meet ANSI Z87.1-2010 and CSA Z94.3 standards.

TRUSTED_SUSTAINABILITY

Specifications

Lens Coating Anti-fog
Lens Type Magnifying
Certifications/Compliance CSA Z94.3 and ANSI Z87.1-2010
Lens Tint Clear
Product Type Reader Protective Eyewear
Magnification Power +2.00
No. per Case 20/Cs.
Model SF420AF
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.