Learn More
3M™ SecureFit™ 500 Series
Features a contemporary lens design coupled with adjustable ratchet temples and 3M™ Pressure Diffusion Temple (PDT) technology to help provide a more personalized fit. This sporty eyewear also offers self-adjusting colored temples, a variety of lens tints and models with Scotchgard™ Protector Anti-Fog Coating.
Supplier:  3M™ 7100239088
 View more versions of this product
Catalog No. 19-072-511
Only null left
Add to Cart
Description
Description
  • 3M's innovative temple design features slotted ribs that provide comfortable pressure equalization, allowing the temples to flex and self-adjust to the size of the wearer's head.
  • Cuved temples apply pressure to the less sensitive back of the head, instead of the temples, achieving a comfortable fit even during rapid head movement.
  • Sporty design with contemporary lens shape and a sleek profile
  • Adjustable ratchet temples allow user to adjust the lens for facial features and eyeglass position.
  • SecureFit's flat and flexible temple design combines well with 3M™ Protective Earmuffs
  • Choose from models with anti-fog/anti-scratch or 3M™ Scotchgard™ Anti-Fog coating.
  • U6 rated polycarbonate lenses absorb 99.9% of UVA, UVB, and UVC rays between 200 nm and 380 nm
  • Workers can feel confident wearing 3M eye protection that meets the Impact-Rated Protector requirement of the ANSI-ISEA Z87.1-2020 Standard
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Anti-fog
5.08 cm
2 in.
Brown
Assembly, Food and Beverage, Military Maintenance, Repair and Operation (MRO), Law Enforcement, Metal Fabrication, Food and Beverage Manufacturing, Chiseling, Demolition, Pharmaceuticals, Primary Metals, Military, Automotive, General Manufacturing, Drilling, Painting, Oil and Gas, Facility Maintenance, Construction, Masonary, Military Tactical, Chemicals, Pouring/Casting, Chemical Handling, Machining, Grinding, Mining, Remediation, Machine Operations, Aviation, Power Fastening
20/Cs.
Polycarbonate
UV400
15.2 cm
Wraparound
6 in.
Protective Eyewear
Olive
SF505SGAF-DGR
Reusable
Frameless
Product Suggestions
Product Suggestions
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.