Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Speedglas Helmet: Battery Holders

Catalog No. 191500512 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
Filters Utility and 9000 Series Welding Helmet
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-150-0512 Filters Utility and 9000 Series Welding Helmet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-150-0512 Supplier 3M™ Supplier No. 7000126764
Add to Cart
Add to Cart

Replacement blue battery holder for 3M™ Speedglas™ Filters Utility and 9000 Series.

  • 3M Catalog# 04-0380-00

TRUSTED_SUSTAINABILITY

Specifications

Color Blue
For Use With (Equipment) Filters Utility and 9000 Series Welding Helmet
Model 04-0380-00
Product Type Battery Holder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.