Learn More
3M™ Speedglas Helmet: Protection Plates
Catalog No. p-4910683 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
Filters 9002X and 9000X/XF/Xi
Fresh-air, PA and SA Helmet
Utility Welding Helmets, Speedglas 9000 Series
Product Type:
Helmet Outside Protection Plate
Inside Protection Plates
Throat and Side Protectors
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Product Type
19-160-0396 Fresh-air, PA and SA Helmet Throat and Side Protectors
19-060-0420 Filters 9002X and 9000X/XF/Xi Inside Protection Plates
19-060-0430 Utility Welding Helmets, Speedglas 9000 Series Helmet Outside Protection Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-160-0396 Supplier 3M™ Supplier No. 7000126769
Only null left
Add to Cart
Add to Cart
Designed for use with Speedglas Helmets
3M offers a variety of replacement protection plates and accessories designed to protect the filter from spark and spatter.
  • Replacement inside protection plates for 3M™ Speedglas™ Filters 9002X and 9000X/XF/Xi
  • Speedglas 9000 standard outside protection plate
  • Replacement silver front panel (only) for 3M™ Speedglas™ FlexView filter assembly
  • Replacement face seal for 3M™ Speedglas™ Helmet 9000 HWR
  • Throat protector with Velcro Fasteners attaches to 3M™ Speedglas™ Helmets 9000 Series
  • Leather throat and side protectors for 3M™ Speedglas™ Fresh-air (FA) PA and SA (APC style helmet)
  • 3M Catalog# 04-0270-01 , 3M Catalog# 04-0280-00 , 3M Catalog# 18-0099-68
Sold separatelyFailure to use an outside protection plate with the filter voids the warranty if spark or spatter contaminates the glass. This option is standard on all Speedglas 9000 Series and Utility Welding Helmets. Outside protection plate made in USA.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Fresh-air, PA and SA Helmet
For Use With (Application) Speedglas Helmet Accessories
Material Leather
Model 18-0099-68
Product Type Throat and Side Protectors