Learn More
3M™ Tri-Flange™ Ear Plugs
Catalog No. p-2676072 Shop All 3M Products
Change view
Click to view available options
Model:
P3000
P3001
Cord Material:
Cloth
Vinyl
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Cord Material
19-130-0470 P3001 Cloth
19-130-0469 P3000 Vinyl
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-130-0470 Supplier 3M™ Supplier No. 7000052724
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Tri-Flange™ flexible flanges adapt to the shape of the ear canal to comfortably reduce exposure to noise. The plastic stem allows the wearer to insert the ear plugs quickly and easily without touching the premolded tips.
  • Triple-flanged, premolded ear plugs fit comfortably in a wide range of sizes
  • Soft, flexible flanges conform to the shape of the ear canal
  • No need to roll ear plugs before fitting so tips stay clean even if hands are dirty
  • Washable and reusable
Compliance
3M™ ear plugs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
TRUSTED_SUSTAINABILITY
Specifications
ANSI S3.1974 Yes
Cord Color Yellow
Cord Material Cloth
CSA Z94.2 Class AL
Size Universal
For Use With (Application) Assembly, Blasting, Cleaning, Composite Finishing, Demolition, Electrical, Facility Maintenance, Final Finish, Grinding, Machine Operations, Paint Preparation, Painting, Sanding, Welding
High Visibility Yes
Model P3001
Packaging Type Poly bag
Pairs per Package 400 Pairs/Cs
Plug Material ABS, elastomeric polymer
Plug Color Clear
Noise Reduction Rating (NRR) 26 dB
Corded Corded
Plug Shape Flanged
Disposable Reusable
Metal Detectable No