Description
- Long, lightweight breathing tube that is easy to wipe down
- Quick release swivel enables one-handed connection and release
- Disposable breathing tube cover is available to help cleaning and decontamination operations
- 3M Catalog# BT-20S and BT-20L
Specifications
Specifications
| Product Type | Breathing Tube |
| Size | Medium/Large |
| For Use With (Application) | Assembly, facility maintenance, grinding, machine operation, painting, sanding |
| For Use With (Equipment) | Versaflo™ PAPR Systems |
| Model | BT-20L |