Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Versaflo™ Healthcare TR-300N+ PAPR Kit

Catalog No. p-7114776 Shop All 3M Products
Change view
Click to view available options
Size:
Medium/Large
Small/Medium
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-000-726 Small/Medium
17-000-727 Medium/Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-000-726 Supplier 3M™ Supplier No. 7100153808
Add to Cart
Add to Cart

Healthcare PAPR Kits are complete, ready-to-use systems with two different size head coverings to choose from. Protects healthcare workers from airborne particulates without compromising comfort or flexibility.

Features:

  • Easy access to ears - Stethoscope use is easier with the unique shape of the S-Series headcovers
  • Connect quickly - Innovative Quick Release Swivel (QRS) connects quickly and helps avoid kinks in the breathing tube
  • Expanded view - See more with a wider field of vision, thanks to enhanced visor design
  • Conveniently disposable - Save time cleaning with the maintenance-free disposable hood
  • Better patient contact - Patients can see their provider’s entire face
  • Two selectable flow rate options offers more options and comfort to the user
  • Easy-to-use and versatile PAPR system for particulate environments
  • High Efficiency particulate filter.
  • Optional nuisance organic vapor filter available
  • Suggested industries include pharmaceutical and healthcare
  • 3M Catalog# R-300N+HKL , 3M Catalog# TR-300N+HKS

Compliance

NIOSH approved.

TRUSTED_SUSTAINABILITY

Specifications

Battery Life 10 to 12 hours
Battery Type High Capacity Lithium Ion
Certifications/Compliance NIOSH
Color Beige
Filter Type HEPA
Product Type PAPR Healthcare Systems and Kits
Size Small/Medium
Flow Rate 6.5 CFM and 7.2 CFM
For Use With (Application) Powder Handling
Model TR-300N+HKS
Mounting Type Belt-mounted
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.