Learn More
3M™ Versaflo™ Uncoated Visor for M-Series PAPR
Catalog No. 19039533 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
M-series PAPR
Catalog No. For Use With (Equipment)
19-039-533 M-series PAPR
1 options
Catalog No. 19-039-533 Supplier 3M™ Supplier No. 7000127690
Add to Cart
Add to Cart
Provides additional molten metal splash protection for M-Series Headtops
  • Provides additional molten metal splash protection for M-Series Headtops
  • 3M Catalog# M-925
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) M-series PAPR
For Use With (Application) Cleaning, electrical, facility maintenance, painting, sanding, welding
Includes One m-921 visor gasket
Material Polycarbonate
Model M-925
Product Type Standard Visor