Learn More
3M™ Versaflo™ TR-300 Series PAPR Accessory, Prefilter
Catalog No. 19039569 Shop All 3M Products
Change view
Click to view available options
Quantity:
10/Pk.
Catalog No. Quantity
19-039-569 10/Pk.
1 options
Catalog No. 19-039-569 Supplier 3M™ Supplier No. 7000127713
Add to Cart
Add to Cart
For installation inside the filter cover of a TR-300 Series PAPR.
  • Prefilter can extend the life of the HE filter by capturing large particles that would otherwise clog the HE filter
  • 3M Catalog# TR-3600
TRUSTED_SUSTAINABILITY
Specifications
Product Type Prefilter
For Use With (Equipment) Versaflo TR-300 PAPR
Model TR-3600
Quantity 10/Pk.