Learn More
Description
- Filter cover helps protect cartridge from physical damage.
- Easy to use cover enables use of prefilters and/or spark arrestors.
- Allows PAPR to be worn through decontamination showers or rain.
- Also used to hold TR-6600 Prefilter or TR-662 Spark Arrestor in place.
- Recommended Industries include healthcare, pharmaceutical, heavy industrial, manufacturing.
Specifications
Specifications
| Certifications/Compliance | NIOSH |
| Color | Gray |
| Product Type | Filter Cover |
| For Use With (Application) | Healthcare, Pharmaceutical |
| For Use With (Equipment) | TR-6500 and TR-600 Respirators, TR-6600 Prefilter, and TR-662 Spark Arrestor |
| Model | TR-6500FC |
| Quantity | 1 Ea. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.