Learn More
Description
- TR-651 filter launch assembly is replacement part for TR-600 motor/blower TR-602N
- Enables simple maintenance of TR-600
- Works with all Versaflo™ TR-6000 series filters and cartridges
- Blue
- 3M Catalog# TR-651
Applications:
Healthcare, Pharmaceutical
Certifications
NIOSH Approved
Specifications
Specifications
| Color | Blue |
| Disposable | Reusable |
| Product Type | Latch Assembly |
| For Use With (Application) | Healthcare, Pharmaceutical |
| For Use With (Equipment) | Versaflo™ TR-600 Series PAPR System |
| Model | TR-651 |
| Quantity | 1 Ea. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.