Learn More
Description
- TR-6600 prefilter extends life of particulate (HE) element of TR-600 series filters and cartridges.
- Captures large particles that may otherwise clog HE filter.
- Frequent prefilter changeout optimizes PAPR system performance.
- Easy to use due to simple maintenance free design.
- Flexible application as TR-6600 may be used with TR-662 spark arrestor.
- Applications include healthcare, pharmaceutical, heavy industrial, manufacturing.
Specifications
Specifications
| Color | Black |
| Product Type | Prefilter |
| For Use With (Application) | Healthcare, Pharmaceutical, Heavy Industrial, Manufacturing, General Manufacturing |
| For Use With (Equipment) | Versaflo™ TR-600 series filters and cartridges |
| Model | TR-6600 |
| Quantity | 50/Cs. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.