Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Virtua AP Protective Eyewear with Hard Coat Lens

Catalog No. 17827320 Shop All 3M Products
Change view
Click to view available options
Certifications/Compliance:
ANSI Z87.1-2010 (U6), CSA Z94.3-2007, & ANSIZ87.1-2010
NSI Z87.1-2010 (U6), CSA Z94.3-2007, & ANSIZ87.1-2010
Lens Tint:
Amber
Blue
Clear
Gray
Indoor/Outdoor Mirror
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance Lens Tint
17-827-320 ANSI Z87.1-2010 (U6), CSA Z94.3-2007, & ANSIZ87.1-2010 Gray
17-827-323 ANSI Z87.1-2010 (U6), CSA Z94.3-2007, & ANSIZ87.1-2010 Clear
17-827-324 ANSI Z87.1-2010 (U6), CSA Z94.3-2007, & ANSIZ87.1-2010 Indoor/Outdoor Mirror
17-827-322 ANSI Z87.1-2010 (U6), CSA Z94.3-2007, & ANSIZ87.1-2010 Amber
17-827-321 NSI Z87.1-2010 (U6), CSA Z94.3-2007, & ANSIZ87.1-2010 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 17-827-320 Supplier 3M™ Supplier No. 7000030050
This item is not returnable. View return policy

Virtua AP protective eyewear offers sleek, unisex styling, lightweight comfort and exceptional value. It is designed with high-wrap polycarbonate lenses and integral side shields to provide comfortable fit and ultimate protection.

3M™ Virtua AP Protective Eyewear is an economy line of safety glasses that offer style, protection and affordability. Providing a wide viewing area, the wraparound design combines with the integrated side shields to keep eyes more fully protected. Contoured temples fit comfortably and snugly without pressure points but also without any looseness that causes shifting. The impact-resistant polycarbonate lens absorbs 99.9% UV rays. These safety glasses are recommended for the following tasks: chipping, chiseling, drilling, grinding, machining, masonry, pouring/casting, power fastening, riveting, sanding and sawing. Industries in which these safety glasses are commonly used include aviation, chemicals, construction, food and beverage, general manufacturing, mining, oil and gas, pharmaceuticals, primary metals, and transportation. Do not use this eyewear in an industrial setting without contacting your supervisor or safety specialist to insure that it will help provide proper protection for the specific hazards in your workplace. Some operations may require special filter lenses, machine guards, goggles, face shields or other safety equipment. Virtua AP safety glasses meet the requirements of CSA Z94.3-2007 and ANSI Z87.1-2010.Eye protection is important to safeguard your eyes and maintain healthy vision. According to the Bureau of Labor Statistics, nearly three out of every five workers who suffered eye injuries were not wearing eye protection at the time of the accident. Wearing the correct safety eyewear for the task and for the unique needs of the worker is important. 3M recognizes the need to design comfortable and attractive eyewear, increasing the usage of safety glasses on the job. That’s why all 3M protective eyewear combines safety features with comfort enhancements.
  • These test earplugs are modified with a probe tube for use during earplug fit testing with the 3M™ E-A-Rfit™ Validation System
  • 3M Catalog# 11815-00000-20, 11816-00000-20, 11817-00000-20, 11818-00000-20, 11847-00000-20

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ANSI Z87.1-2010 (U6), CSA Z94.3-2007, & ANSIZ87.1-2010
Lens Tint Gray
Frame Color Clear
No. per Case 20/Cs.
Model 11815-00000-20
Lens Material Polycarbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.