Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ Virtua AP Protective Eyewear with Hard Coat Lens
Shop All 3M Products
Click to view available options
Certifications/Compliance:
ANSI Z87.1-2010 (U6), CSA Z94.3-2007, & ANSIZ87.1-2010
NSI Z87.1-2010 (U6), CSA Z94.3-2007, & ANSIZ87.1-2010
Lens Tint:
Amber
Blue
Clear
Gray
Indoor/Outdoor Mirror
Description
3M™ Virtua AP Protective Eyewear is an economy line of safety glasses that offer style, protection and affordability. Providing a wide viewing area, the wraparound design combines with the integrated side shields to keep eyes more fully protected. Contoured temples fit comfortably and snugly without pressure points but also without any looseness that causes shifting. The impact-resistant polycarbonate lens absorbs 99.9% UV rays. These safety glasses are recommended for the following tasks: chipping, chiseling, drilling, grinding, machining, masonry, pouring/casting, power fastening, riveting, sanding and sawing. Industries in which these safety glasses are commonly used include aviation, chemicals, construction, food and beverage, general manufacturing, mining, oil and gas, pharmaceuticals, primary metals, and transportation. Do not use this eyewear in an industrial setting without contacting your supervisor or safety specialist to insure that it will help provide proper protection for the specific hazards in your workplace. Some operations may require special filter lenses, machine guards, goggles, face shields or other safety equipment. Virtua AP safety glasses meet the requirements of CSA Z94.3-2007 and ANSI Z87.1-2010.Eye protection is important to safeguard your eyes and maintain healthy vision. According to the Bureau of Labor Statistics, nearly three out of every five workers who suffered eye injuries were not wearing eye protection at the time of the accident. Wearing the correct safety eyewear for the task and for the unique needs of the worker is important. 3M recognizes the need to design comfortable and attractive eyewear, increasing the usage of safety glasses on the job. That’s why all 3M protective eyewear combines safety features with comfort enhancements.
- These test earplugs are modified with a probe tube for use during earplug fit testing with the 3M™ E-A-Rfit™ Validation System
- 3M Catalog# 11815-00000-20, 11816-00000-20, 11817-00000-20, 11818-00000-20, 11847-00000-20

Specifications
Specifications
Certifications/Compliance | NSI Z87.1-2010 (U6), CSA Z94.3-2007, & ANSIZ87.1-2010 |
Lens Tint | Blue |
Frame Color | Clear |
No. per Case | 20/Cs. |
Model | 11816-00000-20 |
Lens Material | Polycarbonate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction