Learn More
3M™ Virtua™ Eyewear
Catalog No. 19121278 Shop All 3M Products
Change view
Click to view available options
Lens Coating:
Anti-fog
Hardcoat
Uncoated
Lens Tint:
Blue Mirror
Clear
Gray
Indoor/Outdoor Mirror
Frame Color:
Blue, mirror
Clear
Gray
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Coating Lens Tint Frame Color
19-121-278 Anti-fog Gray Gray
19-121-277 Anti-fog Clear Clear
19-121-275 Hardcoat Clear Clear
19-121-276 Hardcoat Gray Gray
19-121-279 Hardcoat Indoor/Outdoor Mirror Clear
19-047-295 Uncoated Clear Clear
19-130-0100 - Blue Mirror Blue, mirror
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Catalog No. 19-121-278 Supplier 3M™ Supplier No. 7010386340
Add to Cart
Add to Cart
This item is not returnable. View return policy
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Lightweight, comfortable protection
  • Polycarbonate lens absorbs 99.9% of UV
  • Choice of hardcoat
  • Sleek, 9.5-base lens wraps for superior protection and unobstructed view
  • Contoured flexible temples for comfortable fit
  • Weigh less than one ounce
  • 3M Catalog# 11228-00000-100, 11326-00000-100, 11327-00000-20, 11328-00000-20, 11329-00000-100, 11330-00000-20, 11331-00000-20
Order Info
Weight: <1 oz.
Compliance
Meets ANSI Z87.1-2015 standards.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog
Certifications/Compliance ANSI Z87.1-2015
Lens Tint Gray
Frame Color Gray
Model 11330-00000-20
Lens Material Polycarbonate
Frame Type Contoured, Flexible Temples