Learn More
3M™ Virtua Max Eyewear
Lightweight, unisex Virtua protective eyewear combines versatility and value with high wraparound coverage, polycarbonate construction and a hard coat, scratch-resistant lens for a comfortable, contoured fit.
Specifications
Lens Coating Anti-fog
Lens Material Polycarbonate
View More Specs
Products 1
Catalog Number Mfr. No. Lens Coating Lens Tint Price Quantity & Availability  
Catalog Number Mfr. No. Lens Coating Lens Tint Price Quantity & Availability  
19-810-014
View Documents
3M™
7000127573
Anti-fog Gray N/A N/A N/A
Description
Description
  • Clear anti-fog lens or Indoor/outdoor gray anti-fog lens
  • Clear frame
  • Large lens size
  • Meets the requirements of CSA Z94.3-2007 and the High Impact Requirements of ANSI Z87.1-2003
  • Lightweight, unisex protective eyewear
  • High wraparound coverage
  • Wraparound design provides extended eye protection and unobstructed viewing
  • Contoured temples fit comfortably and snugly
  • Polycarbonate lenses absorb 99.9% UVA and UVB
  • 3M Catalog# 11511-00000-20
Specifications
Specifications
Anti-fog
Polycarbonate
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.