Learn More
3M™ Virtua™ Sport CCS Eyewear
Catalog No. 17827312 Shop All 3M Products
Change view
Click to view available options
Lens Tint:
Clear
Gray
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
17-827-312 Clear
17-827-311 Clear
17-827-313 Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Catalog No. 17-827-312 Supplier 3M™ Supplier No. 7000052872
This item is not returnable. View return policy
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Corded Earplug Control System keeps a corded earplug attached, untangled and ready for use
  • Polycarbonate lenses absorb 99.9% UV
  • Compatible with a variety of vinyl-corded earplugs
  • 3M Catalog# 11796-00000-20 , 3M Catalog# 11797-00000-20 , 3M Catalog# 11798-00000-20
Compliance
Meets ANSI™ Z87. 1-2010 standard.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Meets ANSI Z87. 1-2010 standard.
Lens Tint Clear
No. per Case 20/Cs.
Model 11797-00000-20
Lens Material Polycarbonate