Learn More
3M™ Virtua™ V4 Eyewear
Catalog No. 19161180 Shop All 3M Products
Change view
Click to view available options
Lens Tint:
Clear
Gray
Indoor/Outdoor Mirror
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
19-161-180 Clear
19-161-179 Gray
19-161-182 Indoor/Outdoor Mirror
19-161-181 Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Catalog No. 19-161-180 Supplier 3M™ Supplier No. 7000029998
This item is not returnable. View return policy
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Protection, comfort and style
  • High-impact polycarbonate lens
  • Wraparound lens and unisex styling
  • Soft, dual-injected temples
  • Black temple with gray overmold
  • 3M Catalog# 11674-00000-20, 11673-00000-20, 11672-00000-20, 11671-00000-20, 11670-00000-20
Compliance
Meets ANSI Z87.1-2015 standards.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ANSI Z87.1-2015
Lens Tint Clear
Frame Color Black
Lens Material Polycarbonate