Learn More
Selleck Chemical LLC 3-Methoxytyramine hydrochloride S3370-25mg
SDP
Supplier:  Selleck Chemical LLC S337025MG
Encompass_Preferred
3-Methoxytyramine (3-O-methyl Dopamine 3MT) hydrochloride a major extracellular metabolite of dopamine is a neuromodulator that in certain situations may be involved in movement control
Catalog No. 50-310-8327