Learn More
Apexbio Technology LLC Carbenicillin(Synonyms: Pyopen, Carbenicillin Sodium, Alpha-Carboxybenzylpenicillin, Carboxybenzylpenicillin), 1g, CAS: B3412.
Supplier: Apexbio Technology LLC B34121000
Carbenicillin (CAS 4697-36-3) is a semi-synthetic penicillin derivative characterized by a broad-spectrum antibacterial profile Its bactericidal activity arises primarily through inhibition of bacterial cell wall biosynthesis specifically by interfering with penicillin-binding proteins (PBPs) thus impairing peptidoglycan formation This compound is widely utilized in biomedical research laboratories particularly for selecting bacteria carrying plasmids encoding carbenicillin resistance markers Carbenicillin enables effective differentiation between plasmid-containing and non-plasmid-containing bacterial colonies in molecular biology studies
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.