Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(3S)-(-)-3-Aminopyrrolidine 98.0+%, TCI America™
SDP

Catalog No. A116825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A116825G 25 g
A11685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A116825G Supplier TCI America Supplier No. A116825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 128345-57-3
Molecular Formula C4H10N2
Molecular Weight (g/mol) 86.14
MDL Number MFCD00143193
InChI Key NGXSWUFDCSEIOO-UHFFFAOYNA-N
Synonym s-3-aminopyrrolidine, s-pyrrolidin-3-amine, 3s---3-aminopyrrolidine, 3s-pyrrolidin-3-amine, s---3-aminopyrrolidine, 3-pyrrolidinamine, 3s, 3s-3-aminopyrrolidine, pubchem5729, s-3-pyrrolidinamine, s-3 aminopyrrolidine
PubChem CID 1519351
IUPAC Name pyrrolidin-3-amine
SMILES NC1CCNC1

Specifications

Color Yellow
Quantity 25 g
UN Number 2735
Formula Weight 86.14
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material (3S)-(-)-3-Aminopyrrolidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.