Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(3S)-(-)-3-(Trifluoroacetamido)pyrrolidine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. T13661G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T13661G 1 g
T136625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T13661G Supplier TCI America Supplier No. T13661G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 132883-43-3
Molecular Formula C6H10ClF3N2O
Molecular Weight (g/mol) 218.604
MDL Number MFCD00143195
InChI Key CMZSIQCZAFAEDH-WCCKRBBISA-N
PubChem CID 42614571
IUPAC Name 2,2,2-trifluoro-N-[(3S)-pyrrolidin-3-yl]acetamide;hydrochloride
SMILES C1CNCC1NC(=O)C(F)(F)F.Cl

Specifications

Melting Point 233°C
Color White
Quantity 1 g
Formula Weight 218.60
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (3S)-(-)-3-(Trifluoroacetamido)pyrrolidine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.