Learn More
Medchemexpress LLC (3S,5R)-fluvastatin-d6 sodium | 2249799-35-5 | 98.0% | C24H19D6FNNaO4 | 10MG
SDP
Supplier:  Medchemexpress LLC HY14664CS10MG
Encompass_Preferred
Deuterated (3S,5R)-fluvastatin sodium (d6) is the isotopically labeled sodium salt of the (3S,5R) stereoisomer of fluvastatin. It is supplied as a 10 mg research-grade standard for biochemical, analytical, and pharmacokinetic studies, with documented purity and recommended storage conditions.
  • Deuterated isotopologue for mass spectrometry internal standard applications.
  • High chemical purity suitable for analytical and pharmacological research.
  • Supplied as a sodium salt in a small-mass format for standard preparation.
  • Documented molecular formula and molecular weight for reference.
  • Recommended storage: sealed, away from moisture; in solvent: -80°C (up to 6 months), -20°C (up to 1 month).
Catalog No. 50-002-58957